Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSX1, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002899 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-899 Supplier Abnova Corporation Supplier No. H00004487D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MSX1 protein.

This gene encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. It may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in this gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia. [provided by RefSeq

Sequence: MTSLPLGVKVEDSAFGKPAGGGAGQAPSAAAATAAAMGADEEGAKPKVSPSLLPFSVEALMADHRKPGAKESALAPSEGVQAAGGSAQPLGVPPGSLGAPDAPSSPRPLGHFSVGGLLKLPEDALVKAESPEKPERTPWMQSPRFSPPPARRLSPPACTLRKHKTNRKPRTPFTTAQLLALERKFRQKQYLSIAERAEFSSSLSLTETQVKIWFQNRRAKAKRLQEAELEKLKMAAKPMLPPAAFGLSFPLGGPAAVAAAAGASLYGASGPFQRAALPVAPVGLYTAHVGYSMYHLT

Specifications

Antigen MSX1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MSX1 protein.
Formulation No additive
Gene MSX1
Gene Accession No. NM_002448.2
Gene Alias HOX7/HYD1
Gene Symbols MSX1
Host Species Rabbit
Immunogen MSX1 (AAH67353.1, 1 a.a. ∼ 297 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4487
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.