Learn More
Invivogen Anti-mTLR5-IgG, Neutralizes mTLR5 Activity, Rat Monoclonal Antibody, 100 µg
SDP
Supplier:  Invivogen MABGMTLR52
Encompass
The InvivoGen Anti-mTLR5-IgG is a highly specific rat monoclonal antibody designed for neutralizing murine Toll-like receptor 5 (mTLR5) activity. This reliable tool effectively blocks mTLR5 activation by flagellin, aiding researchers in enhancing their understanding of innate immune responses. It is produced via hybridoma cells and purified through affinity chromatography, ensuring high-quality performance without contamination.
Catalog No. NC2684399