Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mTOR (pS2448) Mouse, PE, Clone: O21-404, BD

Catalog No. BDB563489 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563489 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563489 Supplier BD Biosciences Supplier No. 563489
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The O21-404 monoclonal antibody recognizes the human mammalian target of rapmycin,(mTOR) that is phosphorylated at serine residue 2448, mTOR (pS2448). mTOR belongs to the phosphoinositide-3-kinase (PI3K)-related (PIKK) family of kinases. mTOR is also known as mechanistic target of rapamycin (serine/threonine kinase), FRAP, RAFT1, and RAPT1. mTOR functions as an amino acid and ATP sensor to balance nutrient availability and cell growth. When nutrients are sufficiently available, mTOR is activated by phosphorylation at serine residue 2448 through the PI3 kinase/Akt signaling pathway. Phosphorylated mTOR in turn activates the p70 S6 kinase and contributes to the inactivation 4E-BP1, an elF4E inhibitor. In this way, mTOR participates in the upregulation of cellular protein synthesis, growth, proliferation and survival. mTOR function may be abnormally regulated in tumors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen mTOR (pS2448)
Applications Flow Cytometry
Classification Monoclonal
Clone O21-404
Conjugate PE
Description Mammalian target of rapamycin; FRAP1; RAFT1; RAPT1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.