Learn More
Sartorius Sartorius Anti-Mouse Fc Capture Biosensor, Up To 10 Times Regeneration, Compatible With Octet Systems, Tray
Catalog No. NC0122555 Shop All Sartorius Products
Encompass
The Sartorius Octet Anti-Mouse Fc Capture Biosensors are designed for high-throughput kinetic analysis of mouse Fc-containing proteins. Utilizing Bio-Layer Interferometry (BLI) technology, these biosensors provide a user-friendly solution for studying protein interactions without the need for prior purification. They are regenerable up to 10 times, enabling cost-effective repetitive testing. Each tray contains 96 biosensors, making it ideal for extensive analyses in both crude and purified sample environments.
Catalog No. NC0122555 Supplier Sartorius Supplier No. 185088
The Sartorius Octet Anti-Mouse Fc Capture Biosensors are designed for high-throughput kinetic analysis of mouse Fc-containing proteins. Utilizing Bio-Layer Interferometry (BLI) technology, these biosensors provide a user-friendly solution for studying protein interactions without the need for prior purification. They are regenerable up to 10 times, enabling cost-effective repetitive testing. Each tray contains 96 biosensors, making it ideal for extensive analyses in both crude and purified sample environments.