Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Tag Mouse anti-Human, Clone: 4A6, MilliporeSigma™

Catalog No. 16219MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-219-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-219-MI Supplier MilliporeSigma Supplier No. 16219
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Myc Tag Antibody, clone 4A6, agarose conjugate detects level of Myc Tag and has been published and validated for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myc Tag
Applications Immunoprecipitation
Classification Monoclonal
Clone 4A6
Conjugate Agarose
Formulation Purified mouse monoclonal IgG1 covalently coupled to protein G agarose beads and provided as a 50% slurry in buffer containing PBS, pH 7.4, and 0.05% sodium azide. Liquid suspension.
Gene Accession No. NM_002467.3
Gene Symbols MYC, C-MYC, c-Myc
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.