Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MYH (33-51), Polyclonal

Catalog No. PC709100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC709100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC709100UL Supplier MilliporeSigma™ Supplier No. PC709100UL

Rabbit Polyclonal Antibody

Specifically detects MYH (33-51) Clone: in Human, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin)

Oxidative DNA base lesions such as 8-oxoguanine (8-oxoG) are frequently mutagenic and have been implicated in a variety of pathophysiological states including cancer, Alzheimer's disease, arthritis, and cardiovascular disease. These lesions are generated by oxidative stress. MYH is a mammalian homolog of theE. coli Mut Y DNA glycosylase that excises adenine residues misincorporated opposite the oxidized base, 8-oxoG. MYH up-regulation can be observed following damage in mitochondrial DNA. Single mutations in this protein result in reduced translation efficiency of transcripts. Mutations in MYH have been observed in human colorectal tumors and lung cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH (33-51)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen a synthetic peptide (QEGRQKHAKNNSQAKPSAC) corresponding to amino acids 33-51 of human MYH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the MYH protein and several of its approximately 10 isoforms in PC12, Jurkat, and HeLa cells.
Target Species Human, Rat
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.