Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myogenin Mouse, Unlabeled, Clone: F5D, BD

Catalog No. BDB556358 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556358 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556358 Supplier BD Biosciences Supplier No. 556358
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculin or Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Transfection of myogenin and other family members into a variety of non-muscle cells has been shown to either convert these cells to myogenic cells, or to transcriptionally activate a set of otherwise unexpressed muscle-specific genes. In addition to activating muscle-specific genes, members of the MyoD family members activate their own transcription and transactivate the transcription of other MyoD family members. For example, transfection of myogenin into 10T1/2 cells or Swiss 3T3 cells results in the activation of the endogenous myogenin gene as well as transactivation of MyoD. Likewise, the transfection of MyoD into these cells results in the activation of MyoD as well as the transactivation of myogenin. Each member of the MyoD family has distinct roles in muscle development; myogenin plays a key role in muscle maturation. Myogenin migrates at a molecular weight of ∽34kDa by SDS-PAGE. Clone F5D recognizes human, mouse, rat and cat myogenin. Mapping studies suggest that F5D reacts with an epitope between amino acids 138-158 of myogenin. A recombinant fusion protein corresponding to amino acids 30- 224 of rat myogenin was used as immunogen.

Host Species: Mouse
Clone: F5D
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Rat Myogenin aa. 30-224

Immunohistochemistry, Immunohistochemistry (Frozen), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Myogenin
Applications Western Blot
Classification Monoclonal
Clone F5D
Concentration 0.5mg/mL
Conjugate Unconjugated
Description bHLHc3, Myf-4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Rat Myogenin aa. 30-224
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Feline, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.