Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Myosin Heavy Chain, PE; Clone: MF20; BD

Catalog No. BDB564408 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
PE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
BDB564408 PE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564408 Supplier BD Biosciences Supplier No. 564408
Only null left
Add to Cart
Add to Cart

Applications:

Flow cytometry, Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin Heavy Chain
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MF20
Concentration 0.2 mg/mL
Conjugate PE
Host Species Mouse
Immunogen Chicken pectoralis myosin
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Rat, Zebrafish
Content And Storage Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.