Learn More
Invitrogen™ MYZAP Polyclonal Antibody

Description
A suggested positive control is rat kidney tissue lysate. PA5-21116 can be used with blocking peptide PEP-1230.
Specifications
Specifications
| Antigen | MYZAP |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MYZAP |
| Gene Accession No. | P0CAP1, Q3UIJ9, Q5EB94 |
| Gene Alias | AA407270; AV006038; gcom; GCOM1; Gcom2; Gm640; GRINL1A; GRINL1A combined protein; GRINL1A complex locus 1; GRINL1A complex locus protein 1; GRINL1A complex locus upstream; GRINL1A upstream protein; Grinl1a7; Gup; myocardial intercalated disc protein; myocardial zonula adherens protein; myocardium-enriched ZO1-associated protein; myocardium-enriched zonula adherens protein; myocardium-enriched zonula occludens-1-associated protein; myozap; Myzap; MYZAP-POLR2M |
| Gene Symbols | MYZAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.