Learn More
ABclonal Technology NCS1 Polyclonal Antibody, 100UL, Validated in:WB
Supplier: ABclonal Technology A6455
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human NCS1 (NP_055101.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB•Swiss Prot: P62166•Gene ID:23413•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.