Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nek2 Mouse anti-Human, Unlabeled, Clone: 20, BD

Catalog No. BDB610593 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610593 50 μg
BDB610594 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610593 Supplier BD Biosciences Supplier No. 610593
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reversible protein phosphorylation is critical for progression through the cell cycle and mitosis. In Aspergillus nidulans, the nima gene (never itosis) encodes a protein kinase that is essential for mitosis. Three human genes (nek1, 2, and 3 [Nima-related kinase]) with significant homology to the A. nidulans nima have been reported. The nek2 gene encodes a protein of 445 amino acids and, like its fungal homolog, its expression is regulated throughout the cell cycle. In Hela cells, Nek2 activity and expression are low during M and G1 phases of the cell cycle. However, both parameters increase during S phase and mitosis. In addition, Nek2 phosphorylates protein substrates exclusively at serine and threonine residues. Thus, like its fungal homolog, Nek2 may be a crucial element in controlling the cell's entry into S phase and mitosis.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nek2
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Description Nima Related Kinase 2
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.