Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-Nestin Antibody [RM2043], Rabbit Multiclonal, BSA and Azide Free, 1 mg

Catalog No. 502668758
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-266-8758 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-266-8758 Supplier Abcam Supplier No. AB3160191MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Rabbit multiclonal antibody for Nestin staining in flow cytometry, ICC/IF, and IHC applications.

  • Host species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Applications: IHC-P, ICC/IF, Flow Cyt (Intra), IHC-Fr
  • Reactivity: Human, Mouse, Rat
  • Purification: Affinity purification Protein A
  • Form: Liquid, carrier-free, conjugation-ready
  • Storage buffer: PBS (pH 7.2 to 7.4)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Detection Location In-Cell Detection
Buffer PBS, pH7.2 to 7.4
Form Liquid
Detection Method Chromogenic, Chemiluminescence, Fluorescence
Reactivity Human, Mouse, Rat
Target Molecule Nestin
Label or Dye Unconjugated
Cross Reactivity Human, Mouse, Rat
Formulation PBS, pH 7.2 to 7.4
Quantity 1 mg
For Use With (Application) Flow Cytometry, ICC/IF, IHC-P, IHC-Fr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.