Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuN Guinea Pig anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN90MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN90MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN90MI Supplier MilliporeSigma Supplier No. ABN90
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Detect NeuN using this Anti-NeuN Antibody validated for use in Western Blotting, immunohistochemistry (paraffin), immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuN
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Guinea pig polyclonal serum containing 0.05% sodium azide.
Host Species Guinea Pig
Immunogen GST-tagged recombinant protein corresponding to the N-terminus of mouse NeuN.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.