Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuN Mouse anti-Human, Mouse, Rat, AlexaFluor™488, Clone: A60, MilliporeSigma™

Catalog No. MAB377XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB377XMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB377XMI Supplier MilliporeSigma Supplier No. MAB377X
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NeuN Antibody, clone A60, Alexa Fluor™488 conjugated is an antibody against NeuN for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuN
Applications Immunohistochemistry
Classification Monoclonal
Clone A60
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin conjugated to Alexa Fluor™ 488. Liquid in Phosphate buffer with 15mg/mL BSA as a stabilizer and 0.1% sodium azide.
Gene Symbols NeuN, A60
Host Species Mouse
Immunogen Purified cell nuclei from mouse brain.
Purification Method Protein A purfied
Quantity 500 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.