Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuN Mouse anti-Avian, Chicken, Ferret, Human, Mouse, Pig, Rat, Salamander, Clone: A60, MilliporeSigma™
GSA_VA

Catalog No. MAB377MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB377MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB377MI Supplier MilliporeSigma Supplier No. MAB377
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in flow cytometry, immunocytochemistry, immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuN
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A60
Conjugate Unconjugated
Formulation Purified mouse immunoglobulin IgG1 liquid in buffer containing 0.02Mphosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide.
Gene Symbols NeuN, A58
Host Species Mouse
Immunogen Purified cell nuclei from mouse brain
Purification Method Protein A purified
Quantity 500 μg
Primary or Secondary Primary
Target Species Avian, Chicken, Ferret, Human, Mouse, Pig, Rat, Salamander
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.