Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurocan Mouse anti-Rat, Clone: 650.24, MilliporeSigma™

Catalog No. MAB5212MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5212MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5212MI Supplier MilliporeSigma Supplier No. MAB5212
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Neurocan Antibody, clone 650.24 is validated for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry for the detection of Neurocan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurocan
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 650.24
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_004386.2
Gene Symbols NCAN, FLJ44681, NEUR, CSPG3
Host Species Mouse
Immunogen Embryonic rat brain proteoglycans
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.