Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuroD1 Mouse, PE, Clone: R8-294, BD

Catalog No. BDB563001 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563001 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563001 Supplier BD Biosciences Supplier No. 563001
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The R8-294 monoclonal antibody specifically binds to Neurogenic differentiation factor 1 (NeuroD1). NeuroD1 is a basic helix-loop-helix (bHLH) transcription factor that is also known as BHLHA3, Beta-cell E-box transactivator 2, BETA2 and NeuroD. NeuroD1 interacts with other bHLH proteins and binds the insulin E-box sequence to regulate pancreatic islet cell development. It also plays a key role in central nervous system (CNS) and sensory nervous system neuron development.
Specifically, it is required for the terminal differentiation of neurons during late stages of neurogenesis in the CNS (cerebral cortex, hippocampus, and cerebellum) and the sensory nervous system (eye, inner ear, and olfactory system). During antibody development, the purified R8-294 monoclonal antibody was found to detect NeuroD1 by western blot and indirect immunofluorescent staining followed by flow cytometric analysis or imaging analysis.

Host Species: Mouse
Clone: R8-294
Isotype: IgG1, κ
Species Reactivity: Hu, Ms
Immunogen: Human NeuroD1 Recombinant Protein

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroD1
Applications Flow Cytometry
Classification Monoclonal
Clone R8-294
Conjugate PE
Description Class A basic helix-loop-helix protein 3, bHLHa3, BETA2, NeuroD
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NeuroD1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.