Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide Y Sheep anti-Guinea Pig, Rabbit, Invertebrates, Polyclonal, MilliporeSigma™

Catalog No. AB1583MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1583MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1583MI Supplier MilliporeSigma Supplier No. AB1583
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

Detect Neuropeptide Y using this Anti-Neuropeptide Y Antibody validated for use in immunohistochemistry, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropeptide Y
Applications Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Sheep serum. Lyophilized, no preservatives. Reconstitute with 100μL of sterile distilled water.
Gene Symbols NPY; PYY4
Host Species Sheep
Immunogen Synthetic NPY peptide conjugated to bovine thyroglobulin.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000905.2
Target Species Guinea Pig, Invertebrate, Rabbit
Content And Storage −20-−70°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.