Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Neuropilin-1, Alexa Fluor 700, Clone: 446921, R&D Systems™

Catalog No. FAB3870N100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3870N100 100 Tests
FAB3870N025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB3870N100 Supplier R&D Systems Supplier No. FAB3870N100
This item is not returnable. View return policy

Specifications

Antigen Neuropilin-1
Applications Flow Cytometry
Classification Monoclonal
Clone 446921
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_001019799
Gene Alias BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NRP1, NRPNP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165RCD304
Gene Symbols NRP1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Neuropilin-1, Phe22-Lys644, Accession # NP_001019799
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8829
Test Specificity Detects human Neuropilin-1 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.