Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-κB p65 (pS529) Mouse anti-Human, PE-Cy7, Clone: K10-895.12.50, BD

Catalog No. BDB560335 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560335 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560335 Supplier BD Biosciences Supplier No. 560335
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nuclear factor κB (NF-κB) is a ubiquitously expressed transcription factor that regulates the expression of 200-300 genes. It is crucial for basic cellular responses to stress and pathogens, such as proliferation, survival, development, and apoptosis. The most studied NF-κB complex consists of the p50 (also known as NF-κB1) and p65 (also known as REL-A) subunits, both containing a 300-amino acid region with homology to the Rel proto-oncogene product (RH domain). The RH domain contains motifs for dimerization, nuclear localization, and binding to specific DNA sequences. In addition to the RH domain, the p65 subunit contains the transactivation domain, which is responsible
for the interaction with the inhibitor IκB and which contains phosphorylation sites. In most cell types, the p50/p65 heterodimer is located within the cytoplasm complexed to IκB. This complex prevents nuclear translocation and activity of NF-κB. In response to stimuli such as cytokines, LPS, DNA damage, and viral infections, IκB is phosphorylated at critical residues. This phosphorylation induces dissociation of the IκB/NF-κB complex, allowing the free heterodimeric NF-κB to translocate to the nucleus. Furthermore, optimal activation of NF-κB requires phosphorylation in the transactivation domain of p65. In the nucleus, activated NF-κB dimers bind to the κB sites within promoters and enhancers and function as transcriptional activators.

Host Species: Mouse
Clone: K10-895.12.50
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Phosphorylated Human NF-κB p65 Peptide

Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-κB p65 (pS529)
Applications Flow Cytometry
Classification Monoclonal
Clone K10-895.12.50
Conjugate PE-Cyanine7
Description transcription factor p65, RELA, MGC131774, NFKB3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human NF-κB p65 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.