Learn More
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-NFKB1 (100μg); Mouse purified polyclonal anti-NFKB1 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes a 105kDa protein which can undergo cotranslational processing by the 26S proteasome to produce a 50kDa protein. The 105kDa protein is a Rel protein-specific transcription inhibitor and the 50kDa protein is a DNA binding subunit of the NF-κ-B (NFKB) protein complex. NFKB is a transcription regulator that is activated by various intra- and extra-cellular stimuli such as cytokines, oxidant-free radicals, ultraviolet irradiation, and bacterial or viral products. Activated NFKB translocates into the nucleus and stimulates the expression of genes involved in a wide variety of biological functions. Inappropriate activation of NFKB has been associated with a number of inflammatory diseases while persistent inhibition of NFKB leads to inappropriate immune cell development or delayed cell growth. Two transcript variants encoding different isoforms have been found for this gene.
ELISA
Specifications
Specifications
| Antigen | NFKB1 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NFKB1. |
| Gene | NFKB1 |
| Gene Alias | DKFZp686C01211/EBP-1/KBF1/MGC54151/NF-kappa-B/NFKB-p105/NFKB-p50/p105/p50 |
| Gene Symbols | NFKB1 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.