Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKp46/NCR1 Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4716543
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1850F025 25 Tests
FAB1850F100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1850F025 Supplier R&D Systems Supplier No. FAB1850F025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NKp46/NCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 195314
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Gene Symbols NCR1
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp46/NCR1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9437
Test Specificity Detects human NKp46/NCR1 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.