Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx6.1 Mouse, Alexa Fluor 647, Clone: R11-560, BD

Catalog No. BDB563338 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563338 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563338 Supplier BD Biosciences Supplier No. 563338
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The R11-560 monoclonal antibody specifically binds to human and mouse homeobox protein Nkx6.1. The transcription factor Nkx6.1 plays a role in pancreatic islet beta cell development through the regulation of HNF1a and insulin genes. Nkx6.1 is considered to be a highly specific transcription factor marking pancreatic beta islet cells, where it is expressed in both developing and mature cells. In addition, Nkx6.1 functions as a transcriptional repressor induced by the Sonic Hedgehog (Shh) signaling pathway in fate determination of several neuronal cell types. During antibody development, the purified R11-560 monoclonal antibody was found to detect Nkx6.1 by western blot and indirect immunofluorescent staining followed by flow cytometric analysis or imaging analysis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx6.1
Applications Flow Cytometry
Classification Monoclonal
Clone R11-560
Conjugate Alexa Fluor 647
Description Nkx6.1, NKX6-1, NKX6A, Nkx6-1, Nkx6.1, Nkx6a
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Nkx6.1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.