Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2B Mouse, Unlabeled, Clone: 13, BD

Catalog No. BDB610417 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610417 150 μg
BDB610416 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610417 Supplier BD Biosciences Supplier No. 610417
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The majority of synapses in the central nervous system utilize glutamate as a neurotransmitter to produce rapid neuronal excitation. Glutamate has a diverse array of receptors that can be categorized into two groups: ionotropic and metabotropic. The ionotropic receptors are subdivided into two distint types: 1) receptors for N-methyl D-aspartate (NMDAR) and 2) non-NMDA receptors for AMPA and kainate. Three types of NMDAR2 have been identified: NR2A, NR2B, and NR2C. NR2A and NR2B contain a C-terminal extension (>600 amino acids) that has small scattered regions of conserved sequence. The three NR2 mRNAs show overlapping, differential expression patterns in the rat brain. NR2B has been reported to be expressed in the forebrain, thalamic nuclei, amygdaloid nuclei, caudateputamen, and in restricted regions of the olfactory bulb.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2B
Applications Western Blot
Classification Monoclonal
Clone 13
Concentration 250μg/mL
Conjugate Unconjugated
Description N-Methyl-D-Aspartate Receptor 2B
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.