Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B), Mouse, Clone: 1E4, Abnova™

Catalog No. 89000847 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-847 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-847 Supplier Abnova Corporation Supplier No. H00004882M02
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NPR2.

This gene encodes natriuretic peptide receptor B, one of two integral membrane receptors for natriuretic peptides. Both NPR1 and NPR2 contain five functional domains: an extracellular ligand-binding domain, a single membrane-spanning region, and intracellularly a protein kinase homology domain, a helical hinge region involved in oligomerization, and a carboxyl-terminal guanylyl cyclase catalytic domain. The protein is the primary receptor for C-type natriuretic peptide (CNP), which upon ligand binding exhibits greatly increased guanylyl cyclase activity. Mutations in this gene are the cause of acromesomelic dysplasia Maroteaux type. [provided by RefSeq

Sequence: AKNDHYRTLVRTGPSAPKLGEFVVTLHGHFNWTARAALLYLDARTDDRPHYFTIEGVFEALQGSNLSVQHQVYAREPGGPEQATHFIRANGRIVYICGPL

Specifications

Antigen natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B)
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NPR2.
Formulation PBS with no preservative; pH 7.4
Gene NPR2
Gene Accession No. NM_003995.3
Gene Alias AMDM/ANPRB/GUC2B/GUCY2B/NPRB/NPRBi
Gene Symbols NPR2
Host Species Mouse
Immunogen NPR2 (NP_003986.2, 131 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4882
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.