Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleoporin p62 Mouse, Unlabeled, Clone: 53, BD

Catalog No. p-3850488 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610497 50 μg
BDB610498 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610497 Supplier BD Biosciences Supplier No. 610497
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The nuclear pore complex (NPC) acts as a gate to mediate active transport of proteins and RNA into and out of the nucleus. Proteins actively transported into the nucleus through the NPC require specific nuclear localization sequences. Many of these nucleoporins contain N-acetylglucosamine (GlcNAc) residues that are O-linked to serine or threonine. p62 is the best characterized member of a group of nucleoporins that line the central region of the NPC. A tightly associated complex is formed by p62 and two other nucleoporins, p54 and p58. p54 binds to a carboxy-terminal coiled-coil domain of p62 and p58 binds to a dimer of p54. The amino-terminal domain of p62 contains a series of XFXFX repeats and is joined to the coiled-coil domain by a threonine-rich linker segment. The major role of p62 is maintenance of the structural integrity of NPCs.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleoporin p62
Applications Western Blot
Classification Monoclonal
Clone 53
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.