Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC2 Rat anti-Human, Clone: 3G6, MilliporeSigma™

Catalog No. 05936MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-936-MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-936-MI Supplier MilliporeSigma Supplier No. 05936
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect ORC2 with Anti-ORC2 Antibody, clone 3G6 (Rat Monoclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3G6
Conjugate Unconjugated
Gene Accession No. NP_006181
Gene Symbols ORC2L, ORC2
Host Species Rat
Immunogen Full-length human origin recognition complex 2 (ORC2). Clone 3G6.
Purification Method Protein G purfied
Quantity 50 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.