Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Rex1 Mouse anti-Human, Clone: 6F12, MilliporeSigma™

Catalog No. MABC178MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC178MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC178MI Supplier MilliporeSigma Supplier No. MABC178
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-P-Rex1, clone 6F12 is validated for use in immunoprecipitation, western blot, immunohistochemistry for the detection of P-Rex1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Rex1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6F12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_065871
Gene Symbols PREX1
Host Species Mouse
Immunogen Linear peptide corresponding to human P-Rex 1.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.