Learn More
ABclonal Technology Phospho-MAP3K5-S1033 Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology AP0058
Encompass_Preferred
Immunogen: A synthetic phosphorylated peptide around S1033 of human MAP3K5 (NP_005914.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB•Swiss Prot: Q99683•Gene ID:4217•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 501310092