Learn More
ECM Bioscience p130 Cas (Tyr-751), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP1581
Encompass_Preferred
This antibody was cross-adsorbed to phospho-tyrosine coupled to agarose and to phospho-p130 Cas (Tyr-762) peptide before affinity purification using phospho-p130 Cas (Tyr-751) peptide (without carrier). The antibody detects a 130 kDa* protein corresponding to the molecular mass of phosphorylated p130 Cas on SDS-PAGE immunoblots of pervanadate treated human endothelial and A431 cells, but is not observed in control cells. The immunogen was Phospho-p130 Cas (Tyr-751) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 751 in rat p130 Cas. This rat sequence is the same as Tyr-653 in human p130 Cas and Tyr-657 in mouse p130 Cas. Accession #Q63767. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = BCAR1, Cas
Catalog No. 50-206-8807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.