Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p19 [Skp1] Mouse, Unlabeled, Clone: 52, BD

Catalog No. BDB610530 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610530 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610530 Supplier BD Biosciences Supplier No. 610530

Mouse Monoclonal Antibody

In normal cells, Cyclin A and Cdk2 are components of an active quaternary complex of proteins that includes Cip1 (p21) and PCNA (proliferating cell nuclear antigen). This Cyclin A-Cdk2 complex is required during S phase. In transformed cells, Cyclin A and Cdk2 form complexes with three different proteins: p9, p19, and p45. p19 [Skp1], named for S phase kinase-dependent protein, and p45 [Skp2] are essential elements in the S phase kinase complex that is present in many transformed cell lines. p19 [Skp1] binds to Cyclin A-Cdk2 only when in combination with p45 [Skp2]. The mechanism that mediates p19 and p45 association with Cyclin A-Cdk2 is not yet known.

TRUSTED_SUSTAINABILITY

Specifications

Antigen p19 [Skp1]
Applications Western Blot
Classification Monoclonal
Clone 52
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.