Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p300 CT Mouse anti-Human, Mouse, Rat, Clone: RW128, MilliporeSigma™

Catalog No. 05257MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-257-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-257-MI Supplier MilliporeSigma Supplier No. 05257
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-p300 CT Antibody, clone RW128 (Mouse Monoclonal Antibody) validated in Chimmunoprecipitation, immunoprecipitation, western blotting to detect p300 CT also known as KAT3B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p300 CT
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RW128
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in Protein A Purified immunoglobulin in PBS, pH 7.4, 0.05% sodium azide and 30% glycerol
Gene Accession No. NM_001429
Gene Symbols EP300, p300, P300, OTTHUMP00000028668
Host Species Mouse
Immunogen GST-fusion to the C-Terminus of human p300
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.