Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 (acK382) Mouse anti-Human, Alexa Fluor 647, Clone: L82-51, BD

Catalog No. BDB560231 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560231 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560231 Supplier BD Biosciences Supplier No. 560231
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The p53 protein is critical to regulation of normal cell growth and proliferation and is a suppressor of tumor cell proliferation. Inactivation of p53 by a number of mechanisms, such as missense mutations or interaction with oncogenic viral or cellular proteins, can result in tumor progression. Mutations and/or allelic loss of the p53 gene are associated with a wide variety of human tumors. Known to have a role in transcriptional regulation, p53 suppresses various promoters containing TATA elements in an apparently sequence-independent fashion. p53 also binds to DNA in a sequence-specific manner via recognition of a 20-bp consensus-binding site. This interaction stimulates the expression of genes downstream of the p53 binding site. A number of genes that contain p53-binding sites have been identified, including MDM2, GADD45, and muscle creatine kinase. Post-translational acetylation of p53 enhances its DNA-binding activity. There are multiple factors that affect p53 acetylation, thereby modulating cellular proliferation and apoptosis.
The L82-51 monoclonal antibody recognizes acetylated lysine 382 (acK382) in the C-terminal region of p53.

Host Species: Mouse
Clone: L82-51
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human p53 acetylated Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen p53 (acK382)
Applications Flow Cytometry
Classification Monoclonal
Clone L82-51
Conjugate Alexa Fluor 647
Description TP53 (acK382)
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p53 acetylated Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.