Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Mouse anti-Human, Unlabeled, Clone: PAB 1801, BD

Catalog No. BDB554169 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554169 0.1 mg
BDB554170 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554169 Supplier BD Biosciences Supplier No. 554169
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The gene for the nuclear phosphoprotein p53 is the most commonly mutated gene yet identified in human cancers. Missense mutations occur in tumors of the colon, lung, breast, ovary, bladder and several other organs. The mutant p53 is over-expressed in a variety of transformed cells and it forms specific complexes with several viral oncogenes including SV40 large T, E1B from adenovirus and E6 from human papilloma virus. Recent data suggest that wild type p53 plays a role as a checkpoint protein for DNA damage during the S-phase of the cell cycle. However, it is still unclear whether point mutated forms of p53 are simple null mutants and/or dominant negatively acting proteins. p53 migrates at a reduced molecular weight of 53kDa. Clone PAb 1801 recognizes an epitope between amino acids 32-79 in the N-terminal domain of human wild type and mutant p53 antibody. It does not cross-react with p53 from other species. A truncated recombinant human p53 fusion protein was used as immunogen.

Host Species: Mouse
Clone: PAb 1801
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant fusion protein
Formula Weight [Chemical]: 53kDa

Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Western Blot
Classification Monoclonal
Clone PAB 1801
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant fusion protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.