Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Mouse, Unlabeled, Clone: PAB 240, BD

Catalog No. p-3971659 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554166 0.1 mg
BDB554167 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554166 Supplier BD Biosciences Supplier No. 554166
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p53 is a nuclear phosphoprotein which acts as a tumor suppressor by providing a cell cycle checkpoint for DNA damage during S-phase. Mutations in wildtype p53 can indirectly alter the DNA binding and transcription factor activity of p53. By altering expression of genes normally regulated by p53, these mutations can result in both a loss of tumor suppressor function and a gain of oncogenic function. The majority of mutations in the p53 gene are missense mutations which alter the identity of an amino acid. These mutations may alter the conformation and thus increase the stability of the mutant p53 protein. p53 is expressed in all vertebrate species examined. p53 may be overexpressed in transformed cell lines, where it forms complexes with viral oncogenes including SV40 large T antigen and the adenovirus protein, E1B. p53 migrates at ∽53kDa on SDS-PAGE.
PAb 240 reacts with a conformational epitope between amino acids 156 and 214 of native p53. As such, Pab 240 recognizes only certain mutant forms of p53, as determined by immunoprecipitation. It detects both mutant and wildtype p53 in western blot analysis and immunohistochemistry of frozen tissue sections. It is thought that p53 mutations exert a common conformational change which results in expression of the Pab 240-specific epitope on mutant p53 molecules. A recombinant fusion protein of p53 sequence including amino acids 14-289 was used as immunogen.

Host Species: Mouse
Clone: PAb 240
Isotype: IgG1
Species Reactivity: Human, Mouse, Rat, Hamster, Monkey, Bovine, Chicken
Immunogen: Human p53 aa. 14-289
Formula Weight [Chemical]: 53kDa

Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Western Blot
Classification Monoclonal
Clone PAB 240
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p53 aa. 14-289
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Chicken, Hamster, Human, Monkey, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.