Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Mouse anti-Mouse, Clone: PAb421, MilliporeSigma™

Catalog No. MABE283MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE283MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE283MI Supplier MilliporeSigma Supplier No. MABE283
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-p53 Antibody, clone PAb421 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation, immunohistochemistry to detect p53 also known as Cellular tumor antigen p53, Tumor suppressor p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PAb421
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001120705
Gene Symbols Tp53, P53, Trp53
Host Species Mouse
Immunogen Partially purified mouse p53
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.