Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p54nrb Mouse, Unlabeled, Clone: 3, BD

Catalog No. p-4087615 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611278 50 μg
BDB611279 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611278 Supplier BD Biosciences Supplier No. 611278
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Splicing, the removal of introns from pre-mRNA, is mediated by spliceosomal complexes that contain multiple snRNPs and a number of non-snRNP splicing factors, such as hPRP17 and hPRP18. p54 [nrb] (nuclear RNA-binding protein, 54 kDa) is a non-snRNP RNA splicing factor. It binds and enhances the activity of topoisomerase I, the enzyme that nicks DNA to relieve torsional stress during replication and transcription. p54 [nrb] is the the human homolog of mouse NonO and bovine IPEB. The N-terminal half of p54 [nrb] contains two RNA recognition motifs (RRMs), that allow it to bind single stranded RNA. These motifs are included in a region that is 71% homologous with the human splicing factor PSF and 42% homologous with the Drosophila puff-specific protein BJ6. In addition to its role in RNA splicing, p54 [nrb] functions as a DNA transcription factor in multiple cell types. The N-terminal portion of p54 [nrb] also mediates its interaction with DNA. The ability of p54 [nrb] to bind DNA, but not RNA, is thought to require post-translational modification. Thus, p54 [nrb] can function as either an RNA splicing factor or a DNA transcription factor in a post-translational-dependent manner.

TRUSTED_SUSTAINABILITY

Specifications

Antigen p54nrb
Applications Western Blot
Classification Monoclonal
Clone 3
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p54 [nrb] aa. 368-471
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.