Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p75 LNGFR Mouse anti-Rat, Saporin, Clone: 192, MilliporeSigma™

Catalog No. MAB39025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB39025UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB39025UG Supplier MilliporeSigma Supplier No. MAB39025UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-p75 Low Affinity Nerve Growth Factor Receptor Antibody, Saporin conjugated, clone 192 is validated for use in immunocytochemistry, immunohistochemistry (paraffin), FUNC for the detection of p75 Low Affinity Nerve Growth Factor Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p75 LNGFR
Applications Functional Assay, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 192
Conjugate Saporin
Formulation Saporin Conjugated. 192-IgG-SAP is sterile filtered and presented in Dulbecco's PBS with no preservative.
Gene Accession No. NM_002507.2
Gene Symbols NGFR, p75(NTR), TNFRSF16, Gp80-LNGFR, CD271
Host Species Mouse
Immunogen p75-expressing cells
Purification Method Protein A purfied
Quantity 25 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.