Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAC-1 Mouse anti-Human, Unlabeled, Clone: PAC-1, BD

Catalog No. BDB340535 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340535 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340535 Supplier BD Biosciences Supplier No. 340535
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAC-1 recognizes an epitope on the glycoprotein IIb/IIIa (gpIIb/IIIa, aIIbb3) complex of activated platelets at or near the platelet fibrinogen receptor. The gpIIb/IIIa complex is a member of a family of heterodimeric adhesive protein receptors found on a variety of cell types that have been named the integrin or the cytoadhesion family.The gpIIb/IIIa complex is located on the surface membrane of resting platelets. Platelet activation induces a calcium-dependent conformational change in gpIIb/IIIa that exposes a ligand binding site. Four adhesive macromolecules are capable of interacting with the activated form of gpIIb/IIIa: fibrinogen, von Willebrand factor (vWf), fibronectin, and vitronectin. The binding of fibrinogen to the gpIIb/IIIa receptor is required for platelet aggregation. PAC-1 binds only to activated platelets and appears to be specific for this recognition site within gpIIb/IIIa. Approximately 45,000 to 50,000 gpIIb/IIIa receptors appear on the platelet surface upon activation. PAC-1 inhibits fibrinogen- mediated platelet aggregation.

Flow Cytometry, Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen PAC-1
Applications Flow Cytometry
Classification Monoclonal
Clone PAC-1
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with 0.01% sodium azide.
Host Species Mouse
Immunogen Human platelets
Purification Method Affinity Purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.