Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax-6 Mouse anti-Human, PE, Clone: O18-1330, BD

Catalog No. BDB561552 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561552 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561552 Supplier BD Biosciences Supplier No. 561552
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pax-6 is a member of the paired box (pax) gene family whose protein products are transcription factors involved in development. Pax family members share a highly conserved DNA binding domain that contains six alpha helices (paired domain) and a homeo box domain. Pax-6 has important roles in the development of the eye, nose, central nervous system, and pancreas. Defects in Pax-6 are responsible for various eye malformations including aniridia and Peters anomaly.

The O18-1330 monoclonal antibody reacts with human Pax-6. Because the Pax-6 protein sequence is highly conserved among vertebrate species, cross-reactivity with other species is possible.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax-6
Applications Flow Cytometry
Classification Monoclonal
Clone O18-1330
Conjugate PE
Description Oculorhombin, Aniridia type II protein, PAX6, AN2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Pax-6 aa 406-422 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.