Learn More
Vector Laboratories DuoLuX Substrate, HRP, 200 ml AS-2074
SDP
Supplier:  Vector Laboratories AS20741
Encompass_Preferred
DuoLuX Chemiluminescent and Fluorescent Substrate, Peroxidase (HRP), 200 ml. The substrate can be used for western, southern, northern, or dot blots, or for ELISA applications. This substrate is about 10 times more sensitive than the alkaline phosphatase substrate BCIP/NBT and about 100 times more sensitive than the peroxidase substrate DAB. In addition to its chemiluminescent properties, the reaction product of the DuoLuX Substrate is also fluorescent. Features: Unique formula based on acridan chemistry, Very high sensitivity, Prolonged and intense light emission, Permanent fluorescence. Luminescence: peak emission at 453 nm. Fluorescence: excitation maximum at 405 nm (also excites at 254 nm and 365 nm). Maximum fluorescence emission at 453 nm.
Catalog No. NC1751846
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.