Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphacan Mouse anti-Chicken, MilliporeSigma™

Catalog No. MAB5240MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5240MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5240MI Supplier MilliporeSigma Supplier No. MAB5240
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Phosphacan using this Anti-Phosphacan Antibody validated for use in ELISA, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphacan
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Tissue culture supernatant. Liquid. Contains no preservative.
Gene Symbols PTPRZ1, PTPRZ, PTPZ
Host Species Mouse
Immunogen Mixture of chicken chondroitin sulfate proteoglycans.
Quantity 1 mL
Primary or Secondary Primary
Target Species Chicken
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.