Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-BIK (Thr33) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536706 Supplier Invitrogen™ Supplier No. PA536706
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 23 kDa. Purity is >95% by SDS-PAGE.

The Bik protein is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programmed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-BIK (Thr33)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene BIK
Gene Accession No. Q13323
Gene Alias apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; BCL2 interacting killer; bcl-2-interacting killer; BCL2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; BIK; Bik-like killer protein; Biklk; BIP 1; BIP1; Blk; BP 4; BP4; cb 60; natural born killer; NBK
Gene Symbols BIK
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human BIK/ NBK around the phosphorylation site of Threonine 33.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 638
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.