Learn More
Phospho-BRCA2 (Ser3291) Rabbit anti-Human, Horse, Mouse, Rat, Chimpanzee, Canine, Hamster, Rhesus Macaque, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-BRCA2 (Ser3291) |
| Applications | Dot Blot, Immunocytochemistry, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. |
| Gene Symbols | BRCA2; BRCC2; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to human BRCA2 phosphorylated at and around the internal domain surrounding Ser3291. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.