Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CREB (Ser133) Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06519MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-519-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-519-MI Supplier MilliporeSigma Supplier No. 06519
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-phospho-CREB (Ser133) Antibody (rabbit polyclonal antibody) is highly published and validated in EMSA, immunohistochemistry,immunohistochemistry (paraffin), immunoprecipitation, western blotting to detect phospho-CREB (Ser133) also known as active transcription factor CREB, cAMP responsive element binding protein 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-CREB (Ser133)
Applications Electromobility Shift Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.1M Tris-Glycine (pH 7.4) 15mM NaCl, and 0.05% sodium azide with 30% glycerol.
Gene Symbols CREB1; CREB; MGC9284
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to the amino acids including and encompassing phosphorylated Ser133 of CREB.
Purification Method Purified by protein A-Sepharose followed by immunoadsorbant chromatography using an unphosphorylated CREB affinity gel column.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_134442
Target Species Hamster, Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.