Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p70 S6 Kinase (Thr389) Mouse anti-Human, Mouse, Clone: 10G7.1, MilliporeSigma™

Catalog No. MABS82MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS82MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS82MI Supplier MilliporeSigma Supplier No. MABS82
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-phospho-p70 S6 Kinase (Thr389) Antibody, clone 10G7.1 is validated for use in Western Blotting, immunocytochemistry for the detection of phospho-p70 S6 Kinase (Thr389).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-p70 S6 Kinase (Thr389)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 10G7.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003152
Gene Symbols RPS6KB1, STK14A
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to mouse p70 S6 Kinase phosphorylated at (Thr389).
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.