Learn More
Phospho-α Synuclein (Ser129) Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-α Synuclein (Ser129) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Liquid in 10mM HEPES (pH 7.5), 150mM NaCl, 100 ug/mL BSA and 50% glycerol. |
| Gene Symbols | SNCA; MGC110988; PD1; alpha-synuclein; NACP; PARK1; PARK4; Alpha-synuclein |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide of amino acids surrounding the phosphoSerine 129 site of rat alpha synuclein. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.