Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Ubiquitin-conjugating enzyme E2 A (Ser120) Mouse anti-Mouse, Human, Clone: 7E4, MilliporeSigma™

Catalog No. MABE445MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE445MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE445MI Supplier MilliporeSigma Supplier No. MABE445

Mouse Monoclonal Antibody

Anti-phospho Ubiquitin-conjugating enzyme E2 A (Ser120) Antibody, clone 7E4 is a highly specific mouse monoclonal antibody and that targets Ubiquitin-conjugating enzyme and has been tested in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Ubiquitin-conjugating enzyme E2 A (Ser120)
Applications Western Blot
Classification Monoclonal
Clone 7E4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003327
Gene Symbols UBE2A, RAD6A
Host Species Mouse
Immunogen Linear peptide corresponding to Human phosphorylated Ubiquitin-conjugating enzyme E2 A.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.