Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect RhoA (pSer188), Polyclonal

Catalog No. PS1089100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PS1089100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PS1089100UL Supplier MilliporeSigma™ Supplier No. PS1089100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ RhoA (pSer188) Clone: in Human samples, and it is validated for ELISA, Immuno Blotting

Rho (A, B, and C) proteins are members of the Ras superfamily of GTPases. They regulate a variety of cellular functions, including cell cycle progression, cytoskeletal rearrangement, and gene expression. Rho cycles between the active GTP-bound form and the inactive GDP-bound form. Conversion from the inactive to the active form and vice versa is controlled by guanine nucleotide exchange factors (GEFs) and GTPase-activating proteins (GAPs). The three Rho proteins are highly homologous and contain the consensus sequences necessary for GDP/GTP binding and GTPase activity. RhoA is phosphorylated at Ser188in vitro by PKA and PKG. This phosphorylation enhances RhoGDI binding and inhibits RhoA-mediated stress fiber formation, indicating that Ser188 is an important site for negative regulation of RhoA activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ RhoA (pSer188)
Applications ELISA, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 1mg/mL BSA, 50% glycerol.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids surrounding the Ser188 phosphorylation site of human RhoA, coupled to KLH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽22kDa RhoA protein, expressed as a GST fusion protein (total ∽49kDa) and phosphorylatedin vitro by PKA.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.