Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-epitope SR proteins Mouse anti-Rat, Mouse, Human, Clone: 1H4, MilliporeSigma™

Catalog No. MABE50MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE50MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE50MI Supplier MilliporeSigma Supplier No. MABE50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Use Anti-Phosphoepitope SR proteins Antibody, clone 1H4 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry, immunoprecipitation to detect Phosphoepitope SR proteins also known as Ser-Arg-rich proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoepitope SR proteins
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1H4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols SR proteins
Host Species Mouse
Immunogen Purified oocyte nuclei containing xenopus SR proteins.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.